5-Bromo-2-methoxybenzyl alcohol ≥98.0%

Supplier: TCI America

B3369-25G B3369-5G
TCB3369-25GEA 446.35 CAD
TCB3369-25G TCB3369-5G
5-Bromo-2-methoxybenzyl alcohol ≥98.0%
5-Bromo-2-methoxybenzyl alcohol

CAS Number: 80866-82-6
MDL Number: MFCD00004630
Molecular Formula: C8H9BrO2
Molecular Weight: 217.06
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 73


Other Product Names / Synonyms: 4-Bromo-2-(hydroxymethyl)anisole

Formula: C₈H₉BrO₂
Melting Pt: 73 °C
MDL Number: MFCD00004630
CAS Number: 80866-82-6
EINECS: 279-586-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=063a661dad0962b574881894b5c6b426489120d7&vid=063a661dad0962b574881894b5c6b426489120d7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR