1-Butyl-1-methylpyrrolidinium chloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 1-Butyl-1-methylpyrrolidin-1-ium chloride

B3358-25G B3358-5G
TCB3358-25GEA 254.38 CAD
TCB3358-25G TCB3358-5G
1-Butyl-1-methylpyrrolidinium chloride ≥98.0% (by titrimetric analysis)
1-Butyl-1-methylpyrrolidinium chloride

CAS Number: 479500-35-1
MDL Number: MFCD08458922
Molecular Formula: C9H20ClN
Molecular Weight: 177.72
Purity/Analysis Method: >98.0% (T)
Form: Crystal

Formula: C₉H₂₀ClN
MW: 177.72 g/mol
MDL Number: MFCD08458922
CAS Number: 479500-35-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f71365c80476a2ec1bcdff87ff2b5f17cb2b9305&vid=f71365c80476a2ec1bcdff87ff2b5f17cb2b9305&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR