5-Bromo-2-methoxyphenol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B3342-25G B3342-5G
TCB3342-25GEA 82.16 CAD
TCB3342-25G TCB3342-5G
5-Bromo-2-methoxyphenol ≥98.0% (by GC, titration analysis)
5-Bromo-2-methoxyphenol

CAS Number: 37942-01-1
MDL Number: MFCD00673137
Molecular Formula: C7H7BrO2
Molecular Weight: 203.04
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 150
Melting point (°C): 65


Other Product Names / Synonyms: 4-Bromo-2-hydroxyanisole

Formula: C₇H₇BrO₂
MW: 203.0300 g/mol
Boiling Pt: 150 °C
Melting Pt: 65 °C
MDL Number: MFCD00673137
CAS Number: 37942-01-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e3d57bf15b5750213d4ef3ac378040d46dd7a2a6&vid=e3d57bf15b5750213d4ef3ac378040d46dd7a2a6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR