2-Bromo-3-nitrobenzoic acid ≥98.0%

Supplier: TCI America
Warning

B3322-5G B3322-25G
TCB3322-5GEA 255.96 CAD
TCB3322-5G TCB3322-25G
2-Bromo-3-nitrobenzoic acid ≥98.0%
2-Bromo-3-nitrobenzoic acid

CAS Number: 573-54-6
MDL Number: MFCD00074899
Molecular Formula: C7H4BrNO4
Molecular Weight: 246.02
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 188

Formula: C₇H₄BrNO₄
Melting Pt: 188 °C
MDL Number: MFCD00074899
CAS Number: 573-54-6
EINECS: 209-356-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e98e63de9b8edc09b4b789c9ef705e7170d43644&vid=e98e63de9b8edc09b4b789c9ef705e7170d43644&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR