(3-Bromopropyl)trimethoxysilane ≥98.0%

Supplier: TCI America

Synonyms: Trimethoxy(3-bromopropyl)silane, 3-(Trimethoxysilyl)propylbromide

B3289-5G B3289-25G
TCB3289-5GEA 199.38 CAD
TCB3289-5G 76219-364
(3-Bromopropyl)trimethoxysilane ≥98.0%
(3-Bromopropyl)trimethoxysilane

CAS Number: 51826-90-5
MDL Number: MFCD00053894
Molecular Formula: C6H15BrO3Si
Molecular Weight: 243.17
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 130
Flash Point (°C): 82
Specific Gravity (20/20): 1.30

Formula: BrH₂CCH₂CH₂Si(OCH₃)₃
MW: 243.17 g/mol
Boiling Pt: 130 °C
Density: 1.30 (20 °C)
Flash Pt: 82 °C
MDL Number: MFCD00053894
CAS Number: 51826-90-5
EINECS: 257-453-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0195eda0b83bcec1b789e8bbe4b7fdef874df29d&vid=0195eda0b83bcec1b789e8bbe4b7fdef874df29d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR