3-Bromo-5-fluoropyridine ≥98.0%
Supplier: TCI America
B3244-5G
B3244-1G
TCB3244-5GEA
126.4
CAD
TCB3244-5G
TCB3244-1G
3-Bromo-5-fluoropyridine ≥98.0%
3-Bromo-5-fluoropyridine
CAS Number: 407-20-5
MDL Number: MFCD04112555
Molecular Formula: C5H3BrFN
Molecular Weight: 175.99
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 78
Flash Point (°C): 64
Freezing point (°C): 25
Formula:
C₅H₃BrFN Boiling Pt: 78 °C Flash Pt: 64 °C |
MDL Number:
MFCD04112555 CAS Number: 407-20-5 UN: 2811 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...