2-Bromo-5-chloroaniline ≥97.0%

Supplier: TCI America

B3182-1G B3182-5G B3182-25G
TCB3182-1GEA 26.86 CAD
TCB3182-1G TCB3182-5G TCB3182-25G
2-Bromo-5-chloroaniline ≥97.0%
2-Bromo-5-chloroaniline

CAS Number: 823-57-4
MDL Number: MFCD00672942
Molecular Formula: C6H5BrClN
Molecular Weight: 206.47
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Boiling point (°C): 128
Melting point (°C): 38

Formula: C₆H₅BrClN
Boiling Pt: 128 °C
Melting Pt: 38 °C
MDL Number: MFCD00672942
CAS Number: 823-57-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11ab1ec5bab727bc8ac7a1a4609900da4b8a1dfb&vid=11ab1ec5bab727bc8ac7a1a4609900da4b8a1dfb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR