2-Bromo-4-methoxyphenol ≥98.0% (by titrimetric analysis)

Supplier: TCI America

B3135-25G B3135-5G
TCB3135-25GEA 434.5 CAD
TCB3135-25G TCB3135-5G
2-Bromo-4-methoxyphenol ≥98.0% (by titrimetric analysis)
2-Bromo-4-methoxyphenol

CAS Number: 17332-11-5
MDL Number: MFCD02113712
Molecular Formula: C7H7BrO2
Molecular Weight: 203.04
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 115
Melting point (°C): 43


Other Product Names / Synonyms: 3-Bromo-4-hydroxyanisole

Formula: C₇H₇BrO₂
Boiling Pt: 115 °C
Melting Pt: 43 °C
MDL Number: MFCD02113712
CAS Number: 17332-11-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0006b972d3c05c34ba70f597fc12c5937b52a2f&vid=b0006b972d3c05c34ba70f597fc12c5937b52a2f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR