3-Bromo-4,5-dimethoxybenzaldehyde ≥98.0%

Supplier: TCI America
Warning

Synonyms: 5-Bromoveratraldehyde

B3130-5G B3130-25G
TCB3130-5GEA 229.73 CAD
TCB3130-5G TCB3130-25G
3-Bromo-4,5-dimethoxybenzaldehyde ≥98.0%
3-Bromo-4,5-dimethoxybenzaldehyde

CAS Number: 6948-30-7
MDL Number: MFCD00003346
Molecular Formula: C9H9BrO3
Molecular Weight: 245.07
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 62


Other Product Names / Synonyms: 5-Bromoveratraldehyde

Formula: C₉H₉BrO₃
MW: 245.07 g/mol
Melting Pt: 62 °C
MDL Number: MFCD00003346
CAS Number: 6948-30-7
EINECS: 230-113-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=072486494731c7450c882329f2d17099ca9f604b&vid=072486494731c7450c882329f2d17099ca9f604b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR