3-Bromo-2-naphthol ≥96.0%

Supplier: TCI America

B3127-1G B3127-5G
TCB3127-1GEA 132.72 CAD
TCB3127-1G TCB3127-5G
3-Bromo-2-naphthol ≥96.0%
3-Bromo-2-naphthol

CAS Number: 30478-88-7
Molecular Formula: C10H7BrO
Molecular Weight: 223.07
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 82

Formula: C₁₀H₇BrO
Melting Pt: 82 °C
MDL Number: MFCD10000951
CAS Number: 30478-88-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d1cbe0501554ec840b01347d8aa51f53795bc0f&vid=3d1cbe0501554ec840b01347d8aa51f53795bc0f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR