2-Bromo-4-chloroanisole ≥98.0%
Supplier: TCI America
B3111-25G
B3111-5G
TCB3111-25GEA
115.34
CAD
TCB3111-25G
TCB3111-5G
2-Bromo-4-chloroanisole ≥98.0%
2-Bromo-4-chloroanisole
CAS Number: 60633-25-2
MDL Number: MFCD00079705
Molecular Formula: C7H6BrClO
Molecular Weight: 221.48
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 127
Specific Gravity (20/20): 1.63
Other Product Names / Synonyms: 2-Bromo-4-chloro-1-methoxybenzene
Formula:
C₇H₆BrClO Boiling Pt: 127 °C Density: 1.63 (20 °C) |
MDL Number:
MFCD00079705 CAS Number: 60633-25-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...