2-Bromo-9-fluorenone ≥98.0%

Supplier: TCI America

Synonyms: 2-Bromo-9H-fluoren-9-one

B3109-25G B3109-5G
TCB3109-25GEA 102.7 CAD
TCB3109-25G TCB3109-5G
2-Bromo-9-fluorenone ≥98.0%
2-Bromo-9-fluorenone

CAS Number: 3096-56-8
MDL Number: MFCD00010789
Molecular Formula: C13H7BRO
Molecular Weight: 259.10
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 147

Formula: C₁₃H₇BrO
MW: 259.1 g/mol
Melting Pt: 147 °C
MDL Number: MFCD00010789
CAS Number: 3096-56-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5aecf6ab538c4b6899946e9a4791388d5155a11&vid=e5aecf6ab538c4b6899946e9a4791388d5155a11&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR