1-Butyl-3-methylpyridinium bromide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

B3104-5G B3104-25G
TCB3104-5GEA 134.3 CAD
TCB3104-5G TCB3104-25G
1-Butyl-3-methylpyridinium bromide ≥98.0% (by HPLC, titration analysis)
1-Butyl-3-methylpyridinium bromide

CAS Number: 26576-85-2
MDL Number: MFCD01632121
Molecular Formula: C10H16BrN
Molecular Weight: 230.15
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal

Formula: C₁₀H₁₆BrN
MDL Number: MFCD01632121
CAS Number: 26576-85-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26c34fd6b8535f5d2a67a170cf25cc0cb66084c8&vid=26c34fd6b8535f5d2a67a170cf25cc0cb66084c8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR