1-Butyl-4-nitrobenzene ≥96.0%
Supplier: TCI America
B3092-25G
B3092-5G
TCB3092-25GEA
1407.78
CAD
TCB3092-25G
TCB3092-5G
1-Butyl-4-nitrobenzene ≥96.0%
1-Butyl-4-nitrobenzene
CAS Number: 20651-75-6
Molecular Formula: C10H13NO2
Molecular Weight: 179.22
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 136
Specific Gravity (20/20): 1.06
Formula:
C₁₀H₁₃NO₂ Boiling Pt: 136 °C Density: 1.06 (20 °C) |
CAS Number:
20651-75-6 EINECS: 243-941-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...