1-Butyl-2-nitrobenzene ≥96.0%

Supplier: TCI America

B3091-5G B3091-25G
TCB3091-5GEA 292.62 CAD
TCB3091-5G TCB3091-25G
1-Butyl-2-nitrobenzene ≥96.0%
1-Butyl-2-nitrobenzene

CAS Number: 7137-55-5
Molecular Formula: C10H13NO2
Molecular Weight: 179.22
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 127
Specific Gravity (20/20): 1.06

Formula: C₁₀H₁₃NO₂
Boiling Pt: 127 °C
Density: 1.06 (20 °C)
CAS Number: 7137-55-5
EINECS: 230-435-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b1045b5df3d276f8159d35072ef74919fd70a63&vid=0b1045b5df3d276f8159d35072ef74919fd70a63&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR