5-Bromo-2-fluorophenol ≥98.0% (by GC, titration analysis)
Supplier: TCI America
B3064-5G
B3064-25G
TCB3064-5GEA
61.62
CAD
TCB3064-5G
TCB3064-25G
5-Bromo-2-fluorophenol ≥98.0% (by GC, titration analysis)
5-Bromo-2-fluorophenol
CAS Number: 112204-58-7
MDL Number: MFCD07782066
Molecular Formula: C6H4BrFO
Molecular Weight: 191.00
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 54
Specific Gravity (20/20): 1.76
Formula:
C₆H₄BrFO Boiling Pt: 54 °C Density: 1.76 (20 °C) |
MDL Number:
MFCD07782066 CAS Number: 112204-58-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...