1-Bromo-2-fluoro-4-nitrobenzene ≥95.0%

Supplier: TCI America

Synonyms: 4-Bromo-3-fluoronitrobenzene

B3060-5G B3060-25G
TCB3060-5GEA 80.58 CAD
TCB3060-5G TCB3060-25G
1-Bromo-2-fluoro-4-nitrobenzene ≥95.0%
1-Bromo-2-fluoro-4-nitrobenzene

CAS Number: 185331-69-5
MDL Number: MFCD05865088
Molecular Formula: C6H3BrFNO2
Molecular Weight: 220.00
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 91

Formula: C₆H₃BrFNO₂
Melting Pt: 91 °C
MDL Number: MFCD05865088
CAS Number: 185331-69-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3165217822d443d5c984b630e1482c1081c9b69b&vid=3165217822d443d5c984b630e1482c1081c9b69b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR