5-Bromo-2-fluoroaniline ≥95.0%

Supplier: TCI America

B3043-5G B3043-25G
TCB3043-5GEA 33.18 CAD
TCB3043-5G TCB3043-25G
5-Bromo-2-fluoroaniline ≥95.0%
5-Bromo-2-fluoroaniline

CAS Number: 2924-09-6
MDL Number: MFCD03094181
Molecular Formula: C6H5BrFN
Molecular Weight: 190.02
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 105
Freezing point (°C): 27

Formula: C₆H₅BrFN
Boiling Pt: 105 °C
MDL Number: MFCD03094181
CAS Number: 2924-09-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb194b802474819c52235f4b15085136fa6ead98&vid=eb194b802474819c52235f4b15085136fa6ead98&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR