5-Bromo-2-chloroanisole ≥98.0%

Supplier: TCI America

B3001-5G B3001-1G
TCB3001-5GEA 83.74 CAD
TCB3001-5G TCB3001-1G
5-Bromo-2-chloroanisole ≥98.0%
5-Bromo-2-chloroanisole

CAS Number: 16817-43-9
MDL Number: MFCD00078659
Molecular Formula: C7H6BrClO
Molecular Weight: 221.48
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 130
Flash Point (°C): 113
Freezing point (°C): 26


Other Product Names / Synonyms: 4-Bromo-1-chloro-2-methoxybenzene

Formula: C₇H₆BrClO
Boiling Pt: 130 °C
Flash Pt: 113 °C
MDL Number: MFCD00078659
CAS Number: 16817-43-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe9fb1949ae445829484a0f78cb1309c6bd53cee&vid=fe9fb1949ae445829484a0f78cb1309c6bd53cee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR