2-Bromo-5-chlorobenzaldehyde ≥97.0%

Supplier: TCI America

B2955-25G B2955-5G
TCB2955-25GEA 176.96 CAD
TCB2955-25G TCB2955-5G
2-Bromo-5-chlorobenzaldehyde ≥97.0%
2-Bromo-5-chlorobenzaldehyde

CAS Number: 174265-12-4
Molecular Formula: C7H4BrClO
Molecular Weight: 219.46
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 66

Formula: C₇H₄BrClO
Melting Pt: 66 °C
MDL Number: MFCD00462870
CAS Number: 174265-12-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=858ca2e8785eb9bb78de32c2099343fe0df629d1&vid=858ca2e8785eb9bb78de32c2099343fe0df629d1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR