4-Bromo-3,5-dimethoxybenzaldehyde ≥98.0%
Supplier: TCI America
B2952-5G
B2952-25G
TCB2952-5GEA
173.8
CAD
TCB2952-5G
TCB2952-25G
4-Bromo-3,5-dimethoxybenzaldehyde ≥98.0%
4-Bromo-3,5-dimethoxybenzaldehyde
CAS Number: 31558-40-4
MDL Number: MFCD00040799
Molecular Formula: C9H9BrO3
Molecular Weight: 245.07
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 115
Formula:
C₉H₉BrO₃ Melting Pt: 115 °C |
MDL Number:
MFCD00040799 CAS Number: 31558-40-4 EINECS: 250-698-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...