4-Bromo-2,5-dichlorophenol ≥98.0%

Supplier: TCI America

B2822-25G B2822-500G
TCB2822-25GEA 30.02 CAD
TCB2822-25G TCB2822-500G
4-Bromo-2,5-dichlorophenol ≥98.0%
4-Bromo-2,5-dichlorophenol

CAS Number: 1940-42-7
Molecular Formula: C6H3BrCl2O
Molecular Weight: 241.89
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 187
Melting point (°C): 73

Formula: C₆H₃BrCl₂O
Boiling Pt: 187 °C
Melting Pt: 73 °C
CAS Number: 1940-42-7
EINECS: 217-719-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f15e15f69496aef975dbf5164f8fe72b975c2ea4&vid=f15e15f69496aef975dbf5164f8fe72b975c2ea4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR