2-Bromo-5-chloropyridine ≥98.0%

Supplier: TCI America

B2763-5G B2763-1G
TCB2763-5GEA 60.04 CAD
TCB2763-5G TCB2763-1G
2-Bromo-5-chloropyridine ≥98.0%
2-Bromo-5-chloropyridine

CAS Number: 40473-01-6
Molecular Formula: C5H3BrClN
Molecular Weight: 192.44
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 128
Melting point (°C): 67

Formula: C₅H₃BrClN
Boiling Pt: 128 °C
Melting Pt: 67 °C
MDL Number: MFCD00234006
CAS Number: 40473-01-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=773d83a557320965e6d84e5d1d889fc22a3c8d70&vid=773d83a557320965e6d84e5d1d889fc22a3c8d70&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR