1-Butyl-3-methylimidazolium iodide ≥96.0% (by HPLC, total nitrogen)

Supplier: TCI America

B2708-25G B2708-5G
TCB2708-25GEA 237.96 CAD
TCB2708-25G TCB2708-5G
1-Butyl-3-methylimidazolium iodide ≥96.0% (by HPLC, total nitrogen)
1-Butyl-3-methylimidazolium iodide

CAS Number: 65039-05-6
Molecular Formula: C8H15IN2
Molecular Weight: 266.13
Purity/Analysis Method: >96.0% (HPLC,N)
Form: Clear Liquid

Formula: C₈H₁₅IN₂
Density: 1.49 g/cm³ (20 °C)
Flash Pt: 223 °C
MDL Number: MFCD08276307
CAS Number: 65039-05-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=743784ccae6c2e65885238fa390a5c6a1b9a816c&vid=743784ccae6c2e65885238fa390a5c6a1b9a816c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR