1-Butyl-4-methylpyridinium bromide ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
B2700-25G
B2700-5G
TCB2700-25GEA
456.62
CAD
TCB2700-25G
TCB2700-5G
1-Butyl-4-methylpyridinium bromide ≥98.0% (by HPLC, titration analysis)
1-Butyl-4-methylpyridinium bromide
CAS Number: 65350-59-6
Molecular Formula: C10H16BrN
Molecular Weight: 230.15
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 137
Formula:
C₁₀H₁₆BrN Melting Pt: 137 °C |
MDL Number:
MFCD06798209 CAS Number: 65350-59-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...