2-Bromoanthracene ≥97.0%

Supplier: TCI America

B2616-1G B2616-5G
TCB2616-1GEA 104.28 CAD
TCB2616-1G TCB2616-5G
2-Bromoanthracene ≥97.0%
2-Bromoanthracene

CAS Number: 7321-27-9
Molecular Formula: C14H9Br
Molecular Weight: 257.13
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 220

Formula: C₁₄H₉Br
Melting Pt: 220 °C
MDL Number: MFCD07784002
CAS Number: 7321-27-9

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR