1-Bromoanthracene ≥97.0%

Supplier: TCI America

B2615-500MG B2615-100MG
TCB2615-500MGEA 395 CAD
TCB2615-500MG TCB2615-100MG
1-Bromoanthracene ≥97.0%
1-Bromoanthracene

CAS Number: 7397-92-4
MDL Number: MFCD08276282
Molecular Formula: C14H9Br
Molecular Weight: 257.13
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 100

Formula: C₁₄H₉Br
Melting Pt: 100 °C
MDL Number: MFCD08276282
CAS Number: 7397-92-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11688a605d21fd5a2662e89f85705a17e920d065&vid=11688a605d21fd5a2662e89f85705a17e920d065&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR