1-sec-Butyl-4-nitrobenzene ≥97.0%
Supplier: TCI America
B2428-25G
B2428-5G
TCB2428-25GEA
532.46
CAD
TCB2428-25G
TCB2428-5G
1-sec-Butyl-4-nitrobenzene ≥97.0%
1-sec-Butyl-4-nitrobenzene
CAS Number: 4237-40-5
Molecular Formula: C10H13NO2
Molecular Weight: 179.22
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 144
Specific Gravity (20/20): 1.06
Formula:
C₁₀H₁₃NO₂ Boiling Pt: 144 °C Density: 1.06 (20 °C) |
MDL Number:
MFCD00130025 CAS Number: 4237-40-5 EINECS: 224-196-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...