6-Bromoisatin ≥97.0% (by GC, titration analysis)

Supplier: TCI America

B2424-25G B2424-5G
TCB2424-25GEA 527.72 CAD
TCB2424-25G TCB2424-5G
6-Bromoisatin ≥97.0% (by GC, titration analysis)
6-Bromoisatin

CAS Number: 6326-79-0
Molecular Formula: C8H4BrNO2
Molecular Weight: 226.03
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 274


Product Categories: Synthetic Organic Chemistry, Isatins (Multi-component Reaction)
Other Product Names / Synonyms: 6-Bromoindole-2,3-dione, 6-Bromo-2,3-indolinedione

Formula: C₈H₄BrNO₂
Melting Pt: 274 °C
MDL Number: MFCD01631138
CAS Number: 6326-79-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23e8e410944e0a64c1ab6d54cc55ead12f84d33b&vid=23e8e410944e0a64c1ab6d54cc55ead12f84d33b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR