1-Butyl-4-ethynylbenzene ≥97.0%
Supplier: TCI America
B2301-25G
B2301-5G
TCB2301-25GEA
279.66
CAD
TCB2301-25G
TCB2301-5G
1-Butyl-4-ethynylbenzene ≥97.0%
1-Butyl-4-ethynylbenzene
CAS Number: 79887-09-5
MDL Number: MFCD00173885
Molecular Formula: C12H14
Molecular Weight: 158.24
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Flash Point (°C): 92
Specific Gravity (20/20): 0.90
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: 4-Butylphenylacetylene
Formula:
C₁₂H₁₄ Density: 0.90 (20 °C) Flash Pt: 92 °C |
MDL Number:
MFCD00173885 CAS Number: 79887-09-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...