1,2-Bis(trimethylsilyl)benzene ≥95.0%

Supplier: TCI America

Synonyms: 1,2-Phenylenebis(trimethylsilane)

B2299-1G B2299-5G
TCB2299-1GEA 494.22 CAD
TCB2299-1G TCB2299-5G
1,2-Bis(trimethylsilyl)benzene ≥95.0%
1,2-Bis(trimethylsilyl)benzene

CAS Number: 17151-09-6
MDL Number: MFCD00015590
Molecular Formula: C12H22Si2
Molecular Weight: 222.48
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 0.91
Storage Temperature: 0-10°C

Formula: C₁₂H₂₂Si₂
Density: 0.91 (20 °C)
MDL Number: MFCD00015590
CAS Number: 17151-09-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5da884a80ceef31d51d746166eb10980915a9fc7&vid=5da884a80ceef31d51d746166eb10980915a9fc7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR