1-Butylpyridiniumhexafluorophosphate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

B2196-25G B2196-5G
TCB2196-25GEA 330.22 CAD
TCB2196-25G TCB2196-5G
1-Butylpyridiniumhexafluorophosphate ≥98.0% (by HPLC, titration analysis)
1-Butylpyridinium hexafluorophosphate

CAS Number: 186088-50-6
MDL Number: MFCD03093297
Molecular Formula: C9H14F6NP
Molecular Weight: 281.18
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 75

Formula: C₉H₁₄F₆NP
Melting Pt: 75 °C
MDL Number: MFCD03093297
CAS Number: 186088-50-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6eac703f3e761d70c74eab2239eb7dccd5f1b2fd&vid=6eac703f3e761d70c74eab2239eb7dccd5f1b2fd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR