5'-Bromo-2'-hydroxyacetophenone ≥97.0%

Supplier: TCI America

Synonyms: 1-(5-Bromo-2-hydroxyphenyl)ethan-1-one, 1-(5-Bromo-2-hydroxyphenyl)ethanone

B2061-25G B2061-5G
TCB2061-025GEA 67.78 CAD
TCB2061-025G TCB2061-005G
5'-Bromo-2'-hydroxyacetophenone ≥97.0%
5'-Bromo-2'-hydroxyacetophenone

CAS Number: 1450-75-5
MDL Number: MFCD00191850
Molecular Formula: C8H7BrO2
Molecular Weight: 215.05
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 58


Other Product Names / Synonyms: 2-Acetyl-4-bromophenol

Formula: C₈H₇BrO₂
MW: 215.05
Melting Pt: 58 °C
MDL Number: MFCD00191850
CAS Number: 1450-75-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec11a0bedfb714919cd6156c11eb40dc23d10505&vid=ec11a0bedfb714919cd6156c11eb40dc23d10505&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR