6-Bromoquinoline ≥95.0%
Supplier: TCI America
B2015-1G
B2015-5G
TCB2015-001GEA
36.34
CAD
TCB2015-001G
TCB2015-005G
6-Bromoquinoline ≥95.0%
6-Bromoquinoline
CAS Number: 5332-25-2
MDL Number: MFCD00024023
Molecular Formula: C9H6BrN
Molecular Weight: 208.06
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 116
Freezing point (°C): 19
Specific Gravity (20/20): 1.55
Storage Temperature: 0-10°C
Formula:
C₉H₆BrN Boiling Pt: 116 °C Density: 1.55 (20 °C) |
MDL Number:
MFCD00024023 CAS Number: 5332-25-2 EINECS: 226-238-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...