5-Bromo-2,4-dimethoxypyrimidine ≥98.0%

Supplier: TCI America

B1920-25G B1920-1G B1920-5G
TCB1920-25GEA 851.62 CAD
TCB1920-25G TCB1920-001G TCB1920-005G
5-Bromo-2,4-dimethoxypyrimidine ≥98.0%
5-Bromo-2,4-dimethoxypyrimidine

CAS Number: 56686-16-9
MDL Number: MFCD00038016
Molecular Formula: C6H7BrN2O2
Molecular Weight: 219.04
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 125
Melting point (°C): 63

Formula: C₆H₇BrN₂O₂
MW: 219.0400 g/mol
Boiling Pt: 125 °C
Melting Pt: 63 °C
MDL Number: MFCD00038016
CAS Number: 56686-16-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c0696e68f571de12270f1c61021440fd30d8637&vid=4c0696e68f571de12270f1c61021440fd30d8637&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR