1-Bromo-3-chloro-2-methylpropane ≥98.0%
Supplier: TCI America
|
Warning
|
B1859-25G
B1859-500G
TCB1859-025GEA
56.25
CAD
TCB1859-025G
TCB1859-500G
1-Bromo-3-chloro-2-methylpropane ≥98.0%
1-Bromo-3-chloro-2-methylpropane
CAS Number: 6974-77-2
MDL Number: MFCD00000955
Molecular Formula: C4H8BrCl
Molecular Weight: 171.46
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 154
Flash Point (°C): 110
Specific Gravity (20/20): 1.49
Formula:
C₄H₈BrCl Boiling Pt: 154 °C Density: 1.49 (20 °C) Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000955 CAS Number: 6974-77-2 EINECS: 230-224-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...