7-Bromo-1-heptanol ≥88.0%

Supplier: TCI America

B1852-1G B1852-5G
TCB1852-001GEA 90.06 CAD
TCB1852-001G TCB1852-005G
7-Bromo-1-heptanol ≥88.0%
7-Bromo-1-heptanol

CAS Number: 10160-24-4
MDL Number: MFCD00041708
Molecular Formula: C7H15BrO
Molecular Weight: 195.10
Purity/Analysis Method: >88.0% (GC)
Form: Clear Liquid
Flash Point (°C): 110
Specific Gravity (20/20): 1.28
Storage Temperature: <0°C

Formula: C₇H₁₅BrO
MW: 195.10 g/mol
Density: 1.28 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00041708
CAS Number: 10160-24-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c6db3e04c845cafb4676bf3549c20db9403c7ec3&vid=c6db3e04c845cafb4676bf3549c20db9403c7ec3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 88.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR