5-Bromo-1-pentanol ≥90.0%

Supplier: TCI America

B1848-5G B1848-25G
TCB1848-005GEA 71.86 CAD
TCB1848-005G TCB1848-025G
5-Bromo-1-pentanol ≥90.0%
5-Bromo-1-pentanol

CAS Number: 34626-51-2
MDL Number: MFCD00061101
Molecular Formula: C5H11BrO
Molecular Weight: 167.05
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 117
Specific Gravity (20/20): 1.38
Storage Temperature: <0°C

Formula: C₅H₁₁BrO
Boiling Pt: 117 °C
Density: 1.38 (20 °C)
MDL Number: MFCD00061101
CAS Number: 34626-51-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4bd1aabddac1df5ad62b00986e433c45ddbecdbd&vid=4bd1aabddac1df5ad62b00986e433c45ddbecdbd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR