1-Bromo-3,5-dimethyladamantane ≥98.0%

Supplier: TCI America

B1821-5G B1821-25G
TCB1821-5GEA 82.16 CAD
TCB1821-5G TCB1821-025G
1-Bromo-3,5-dimethyladamantane ≥98.0%
1-Bromo-3,5-dimethyladamantane

CAS Number: 941-37-7
MDL Number: MFCD00077197
Molecular Formula: C12H19Br
Molecular Weight: 243.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 273
Flash Point (°C): 109
Specific Gravity (20/20): 1.24

Formula: C₁₂H₁₉Br
Boiling Pt: 273 °C
Density: 1.24 (20 °C)
Flash Pt: 109 °C
MDL Number: MFCD00077197
CAS Number: 941-37-7
EINECS: 213-378-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0fcbb48bef37efb7afd25ab51225d152d4cb6ac7&vid=0fcbb48bef37efb7afd25ab51225d152d4cb6ac7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR