9-Bromo-1-nonanol ≥95.0%
Supplier: TCI America
B1785-25G
B1785-5G
TCB1785-025GEA
169.06
CAD
TCB1785-025G
TCB1785-5G
9-Bromo-1-nonanol ≥95.0%
9-Bromo-1-nonanol
CAS Number: 55362-80-6
MDL Number: MFCD00063348
Molecular Formula: C9H19BrO
Molecular Weight: 223.15
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 126
Flash Point (°C): 109
Freezing point (°C): 30
Storage Temperature: 0-10°C
Formula:
C₉H₁₉BrO Boiling Pt: 126 °C Flash Pt: 109 °C |
MDL Number:
MFCD00063348 CAS Number: 55362-80-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...