12-Bromo-1-dodecanol ≥98.0%

Supplier: TCI America

B1731-25G B1731-5G
TCB1731-25GEA 775.78 CAD
TCB1731-25G TCB1731-005G
12-Bromo-1-dodecanol ≥98.0%
12-Bromo-1-dodecanol

CAS Number: 3344-77-2
MDL Number: MFCD00004754
Molecular Formula: C12H25BrO
Molecular Weight: 265.24
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 156
Melting point (°C): 33
Flash Point (°C): 110
Storage Temperature: 0-10°C

Formula: HO(CH₂)₁₁CH₂Br
Boiling Pt: 156 °C
Melting Pt: 33 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004754
CAS Number: 3344-77-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=19791638e9dcad6b238ac3eb96934f12f2167cb8&vid=19791638e9dcad6b238ac3eb96934f12f2167cb8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR