Bromohydroquinone ≥90.0%

Supplier: TCI America

B1600-25G B1600-5G
TCB1600-25GEA 821.6 CAD
TCB1600-25G TCB1600-005G
Bromohydroquinone ≥90.0%
Bromohydroquinone

CAS Number: 583-69-7
MDL Number: MFCD00041747
Molecular Formula: C6H5BrO2
Molecular Weight: 189.01
Purity/Analysis Method: >90.0% (GC)
Form: Crystal


Other Product Names / Synonyms: 2-Bromo-1,4-benzenediol, Bromoquinol

Formula: C₆H₅BrO₂
MDL Number: MFCD00041747
CAS Number: 583-69-7
EINECS: 209-516-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1f47a1c33871eb17687c7d47e3846b5576c88f29&vid=1f47a1c33871eb17687c7d47e3846b5576c88f29&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR