3-Bromo-4-methylthiophene ≥93.0%

Supplier: TCI America

B1541-25G B1541-5G
TCB1541-25GEA 423.44 CAD
TCB1541-25G TCB1541-005G
3-Bromo-4-methylthiophene ≥93.0%
3-Bromo-4-methylthiophene

CAS Number: 30318-99-1
MDL Number: MFCD00130084
Molecular Formula: C5H5BrS
Molecular Weight: 177.06
Purity/Analysis Method: >93.0% (GC)
Form: Clear Liquid
Boiling point (°C): 180
Flash Point (°C): 77
Specific Gravity (20/20): 1.60

Formula: C₅H₅BrS
Boiling Pt: 180 °C
Density: 1.60 (20 °C)
Flash Pt: 77 °C
MDL Number: MFCD00130084
CAS Number: 30318-99-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7cea2bfcd740b94b6674e62ed6edb8cb04a10f32&vid=7cea2bfcd740b94b6674e62ed6edb8cb04a10f32&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR