1,4-Bis(trimethylsilyl)-1,3-butadiyne ≥99.0%

Supplier: TCI America

Synonyms: 1,4-Bis(trimethylsilyl)buta-1,3-diyne, 1,4-Bis(trimethylsilyl)butadiyne

B1298-1G B1298-5G
TCB1298-001GEA 147.34 CAD
TCB1298-001G TCB1298-005G
1,4-Bis(trimethylsilyl)-1,3-butadiyne ≥99.0%
1,4-Bis(trimethylsilyl)-1,3-butadiyne

CAS Number: 4526-07-2
MDL Number: MFCD00009839
Molecular Formula: C10H18Si2
Molecular Weight: 194.42
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 112

Formula: C₁₀H₁₈Si₂
MW: 194.42 g/mol
Melting Pt: 112 °C
MDL Number: MFCD00009839
CAS Number: 4526-07-2
UN: 1325
ADR: 4.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3ee56c1771215c50f194b2e780159df73c55871&vid=a3ee56c1771215c50f194b2e780159df73c55871&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR