5-Bromoacenaphthene ≥93.0%

Supplier: TCI America

Synonyms: 1,2-Dihydro-5-bromoacenaphthylene, 5-Bromo-1,2-dihydroacenaphthylene

B1283-5G B1283-25G
TCB1283-005GEA 47.4 CAD
TCB1283-005G TCB1283-25G
5-Bromoacenaphthene ≥93.0%
5-Bromoacenaphthene

CAS Number: 2051-98-1
MDL Number: MFCD00003809
Molecular Formula: C12H9Br
Molecular Weight: 233.11
Purity/Analysis Method: >93.0% (GC)
Form: Crystal
Boiling point (°C): 335
Melting point (°C): 55
Flash Point (°C): 110

Formula: C₁₂H₉Br
MW: 233.11 g/mol
Boiling Pt: 335 °C
Melting Pt: 55 °C
Density: 1.1452 g/cm³ (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00003809
CAS Number: 2051-98-1
EINECS: 218-138-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7574e83f800776b37b73e932372018de1376a2c8&vid=7574e83f800776b37b73e932372018de1376a2c8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR