(Bromomethyl)triphenylphosphonium bromide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

B1206-5G B1206-25G
TCB1206-005GEA 85.21 CAD
TCB1206-005G TCB1206-025G
(Bromomethyl)triphenylphosphonium bromide ≥98.0% (by titrimetric analysis)
(Bromomethyl)triphenylphosphonium bromide

CAS Number: 1034-49-7
MDL Number: MFCD00011864
Molecular Formula: C19H17Br2P
Molecular Weight: 436.13
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 243

Formula: C₁₉H₁₇Br₂P
Melting Pt: 243 °C
MDL Number: MFCD00011864
CAS Number: 1034-49-7
EINECS: 213-857-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75f53c66ebf343d5413b4bdedaf3246749b60fc8&vid=75f53c66ebf343d5413b4bdedaf3246749b60fc8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR