2,2-Bis(hydroxymethyl)propionic acid ≥97.0% (by titrimetric analysis)

Supplier: TCI America

B1192-25G B1192-500G
TCB1192-025GEA 31.6 CAD
TCB1192-025G TCB1192-500G
2,2-Bis(hydroxymethyl)propionic acid ≥97.0% (by titrimetric analysis)
2,2-Bis(hydroxymethyl)propionic acid

CAS Number: 4767-03-7
MDL Number: MFCD00004199
Molecular Formula: C5H10O4
Molecular Weight: 134.13
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Boiling point (°C): 200
Melting point (°C): 185
Flash Point (°C): 220


Other Product Names / Synonyms: Dimethylolpropionic Acid, Bis-MPA

Formula: C₅H₁₀O₄
Boiling Pt: 200 °C
Melting Pt: 185 °C
Flash Pt: 220 °C
MDL Number: MFCD00004199
CAS Number: 4767-03-7
EINECS: 225-306-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0004a5d336a9c664442eeab33c0677777a294ac8&vid=0004a5d336a9c664442eeab33c0677777a294ac8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR