(S)-(+)-1,3-Butanediol ≥98.0%

Supplier: TCI America

B1160-1G B1160-5G
TCB1160-001GEA 91.64 CAD
TCB1160-001G TCB1160-5G
(S)-(+)-1,3-Butanediol ≥98.0%
(S)-(+)-1,3-Butanediol

CAS Number: 24621-61-2
MDL Number: MFCD00064278
Molecular Formula: C4H10O2
Molecular Weight: 90.12
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 109
Flash Point (°C): 122
Specific Gravity (20/20): 1.01
Specific rotation [a]20/D: 30 deg (C=1, EtOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-(+)-1,3-Butylene Glycol, (S)-(+)-1,3-Dihydroxybutane

Formula: C₄H₁₀O₂
Boiling Pt: 109 °C
Density: 1.01 (20 °C)
Flash Pt: 122 °C
MDL Number: MFCD00064278
CAS Number: 24621-61-2
EINECS: 246-363-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7023804dc5e4281311f8eb04df4255a704020dfe&vid=7023804dc5e4281311f8eb04df4255a704020dfe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR