(R)-(-)-1,3-Butanediol ≥98.0%

Supplier: TCI America

Synonyms: (R)-(-)-1,3-Dihydroxy butane, (R)-(-)-1,3-Butylene glykol, (R)-Butane-1,3-diol

B1159-1G B1159-5G
TCB1159-001GEA 85.21 CAD
TCB1159-001G TCB1159-5G
(R)-(-)-1,3-Butanediol ≥98.0%
(R)-(-)-1,3-Butanediol

CAS Number: 6290-03-5
MDL Number: MFCD00064277
Molecular Formula: C4H10O2
Molecular Weight: 90.12
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 117
Flash Point (°C): 122
Specific Gravity (20/20): 1.01
Specific rotation [a]20/D: -30 deg (C=1, EtOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-(-)-1,3-Butylene Glycol, (R)-(-)-1,3-Dihydroxybutane

Formula: C₄H₁₀O₂
MW: 90.12 g/mol
Boiling Pt: 117 °C
Density: 1.01 (20 °C)
Flash Pt: 122 °C
MDL Number: MFCD00064277
CAS Number: 6290-03-5
EINECS: 228-532-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1e5d0555a57f25db95670c7be6f77a12ebe8b1a&vid=e1e5d0555a57f25db95670c7be6f77a12ebe8b1a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR