1,2-Bis(diphenylphosphino)ethane ≥97.0%

Supplier: TCI America
Warning

Synonyms: Ethylenebis(diphenylphosphine), Bis(1,2-diphenylphosphino)ethane, DIPHOS, 1,2-Bis(diphenylphosphino)ethane, Ethane-1,3-diylbis(diphenylphosphine)

B1137-10G B1137-25G
TCB1137-010GEA 63.47 CAD
TCB1137-010G TCB1137-25G
1,2-Bis(diphenylphosphino)ethane ≥97.0%
1,2-Bis(diphenylphosphino)ethane

CAS Number: 1663-45-2
MDL Number: MFCD00003047
Molecular Formula: C26H24P2
Molecular Weight: 398.43
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 142


Product Categories: Synthetic Organic Chemistry, Cross-coupling Reaction using Transition Metal Catalysts, Ligands, Ligands ([2+2+2]Cycloaddition Reaction)
Other Product Names / Synonyms: Ethylenebis(diphenylphosphine)

Formula: C₂₆H₂₄P₂
MW: 398.42 g/mol
Melting Pt: 142 °C
Storage Temperature: Ambient
MDL Number: MFCD00003047
CAS Number: 1663-45-2
EINECS: 216-769-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=859b79caed6a986beb95f628fa03feeffd7ed0a1&vid=859b79caed6a986beb95f628fa03feeffd7ed0a1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR